Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Antibody (817G7), Novus Biologicals™
SDP

Catalog No. DDX0377P100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0377P100 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0377P100 Supplier Novus Biologicals Supplier No. DDX0377P100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

Langerin/CD207 Monoclonal specifically detects Langerin/CD207 in Human samples. It is validated for Flow Cytometry, Functional Assay, In-Cell Western (ICW), Inhibition Activity.

Specifications

Antigen Langerin/CD207
Applications Flow Cytometry, Functional Assay, In-cell Western (ICW), Inhibition Assays
Classification Monoclonal
Clone 817G7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Functional, Inhibition Activity
Gene Alias CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Gene Symbols CD207
Host Species Mouse
Immunogen human Langerin-transfected eukaryotic cells
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50489
Test Specificity human Langerin (extracellular epitope)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.