Learn More
Description
Specifications
Specifications
| Antigen | Langerin/CD207 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 923B7 |
| Concentration | 0.5mg/mL |
| Conjugate | Alexa Fluor 546 |
| Dilution | Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Gene Accession No. | NP_056532 |
| Gene Alias | CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin |
| Gene Symbols | CD207 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
