Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Antibody (LGRN/3136R), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB389889 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389889 Supplier Novus Biologicals Supplier No. NBP308872R
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for ELISA

Specifications

Antigen Langerin/CD207
Applications ELISA
Classification Monoclonal
Clone LGRN/3136R
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Host Species Rabbit
Immunogen Recombinant fragment (around aa74-213) of human Langerin/CD207 protein (exact sequence is proprietary) (Uniprot: Q9UJ71)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50489
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.