Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Langerin/CD207 Antibody (LGRN/7430), Novus Biologicals™
SDP

Catalog No. NB233466 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB233466 20 μg
NB232481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB233466 Supplier Novus Biologicals Supplier No. NBP32055620UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Langerin/CD207
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LGRN/7430
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Host Species Mouse
Immunogen Recombinant fragment (around aa50-250) of human Langerin/CD207 protein (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50489
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.