Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Langerin/CD207 Antibody (LGRN/7430), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | Langerin/CD207 |
---|---|
Clone | LGRN/7430 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Immunohistochemistry (Paraffin) |
Description
Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Langerin/CD207 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
metabolism, Signal Transduction | |
10mM PBS with 0.05% BSA | |
50489 | |
IgG | |
Protein A or G purified |
LGRN/7430 | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin | |
Recombinant fragment (around aa50-250) of human Langerin/CD207 protein (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title