Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LANPL Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP177227 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177227 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177227 Supplier Novus Biologicals Supplier No. NBP177227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LANPL Polyclonal specifically detects LANPL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.

Specifications

Antigen LANPL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 2 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. NP_112182
Gene Alias acidic (leucine-rich) nuclear phosphoprotein 32 family, member E, LANP-like protein, LANP-LLANPLacidic leucine-rich nuclear phosphoprotein 32 family member E, leucine-rich acidic nuclear protein like, MGC5350
Gene Symbols ANP32E
Host Species Rabbit
Immunogen Antibody was raised with a synthetic peptide corresponding to amino acids close to carboxy terminus of human PHAP III. The immunogen is located within amino acids 200 - 250 of PHAP III. Amino Acid Squence: GLSYLMKEEIQDEE
Molecular Weight of Antigen 35 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 81611
Test Specificity PHAP III has no cross-reaction to PHAP I and PHAP I2a.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.