Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Catalog No. AA1127330
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1127330 250 g
AA1127318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1127330 Supplier Thermo Scientific Chemicals Supplier No. 01127330
Only null left
Add to Cart

CAS: 13709-38-1 | F3La | 195.90 g/mol

Lanthanum(III) fluoride is used in phosphor lamp coating, carbon arc electrodes and in lasers. It is also used in fluoride selective electrodes. Further, it is used as a non-ferrous metal additive and used in metal alloys.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-38-1
Molecular Formula F3La
Molecular Weight (g/mol) 195.90
MDL Number MFCD00011073
InChI Key BYMUNNMMXKDFEZ-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trifluoride
SMILES [F-].[F-].[F-].[La+3]

Specifications

Melting Point 1493°C
Color White
Physical Form Crystals
Quantity 250 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in strong mineral acids. Insoluble in water.
Formula Weight 195.9
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 5.936 g/cm3 at 20°C
Chemical Name or Material Lanthanum(III) fluoride, anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-252-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.