Learn More
Lanthanum(III) isopropoxide, La 40%
Description
Lanthanum(III) isopropoxide finds application in asymmetric catalysis. It forms complex with an (S,S)-phenyl-linked-binaphthol ,which enhances a catalytic 1,4-addition of malonate to cyclic and acyclic enones. It acts as a co-catalyst with copper fluoride-triphenylphosphine in catalytic allylation of ketoimines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 120°C to 128°C |
| Color | White |
| Boiling Point | 170°C (0.04 mmHg)(sublimation) |
| Odor | Alcohol-like |
| Linear Formula | La[OCH(CH3)2]3 |
| Quantity | 2 g |
| UN Number | UN2925 |
| Sensitivity | Air and moisture sensitive |
| Solubility Information | Soluble in terahydrofuran,toluene,hexane and isopropyl alcohol. |
| Formula Weight | 316.18 |
| Show More |
Safety and Handling
| Hazard Category | H228-H261-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H228 Causes severe skin burns and eye damage. Causes serious eye damage. Flammable solid. |
| Precautionary Statement | P210-P223-P231+P232-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c |
| DOTInformation | Hazard Class: 4.1; Packaging Group: II |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.