Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Oxide, Reagents

Catalog No. 01337201
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-201 100 g
01-337-200 10 kg
01-337-199 1 kg
01-337-198 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-201 Supplier Reagents Holdings Llc Supplier No. C2224350100C2
Only null left
Add to Cart
Add to Cart

Lanthanum Oxide, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1312-81-8
Molecular Formula La2O3
Molecular Weight (g/mol) 325.81
InChI Key MRELNEQAGSRDBK-UHFFFAOYSA-N
Synonym Lanthanum sesqioxide
IUPAC Name dilanthanum(3+) trioxidandiide
SMILES [O--].[O--].[O--].[La+3].[La+3]

Specifications

Boiling Point 4200°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 6.51
Chemical Name or Material Lanthanum Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.