Learn More
Thermo Scientific™ LanthaScreen™ PDCD4 HEK 293E Cell Line
Description
The PI3K/AKT/mTOR pathway mediates signals for cell growth and survival, transcription and translation, and regulated cell death. The significance of mTOR has been underscored recently by the identification of two distinct cellular complexes: mTORC1 (includes raptor and is rapamycin sensitive) and mTORC2 (includes rictor and is insensitive to rapamycin). Activation of the pathway by insulin (or other mitogens) leads to the phosphorylation of programmed cell death protein 4 (PDCD4) via mTORC1 (indirectly through the ribosomal protein p70 S6 kinase). LanthaScreen™ PDCD4 HEK293E is a human cell line which constitutively expresses GFP-PDCD4 fusion proteins. This kinase target was introduced using lipid transfection followed by selection with Blasticidin. These cells are a clonal population isolated by FACS using GFP fluorescence as a sorting marker. A homogenous immunoassay ('addition only', no wash steps) was developed in 384-well format using a TR FRET-based detection method to monitor the phosphorylation of Ser457 on GFP-PDCD4 using a terbium-labeled phosphospecific antibody. The assay has been optimized for performance under numerous experimental conditions (e.g., cell plating density, DMSO tolerance, insulin stimulation time, etc.) and displays excellent statistical data (Z' > 0.6). Moreover, this cell line has been validated with different stimuli and known inhibitors (shows correct EC50 and IC50 values) and serves as robust cellular readout for mTORC1 signaling.
Specifications
Specifications
| Alternate Agonists | Insulin |
| Antagonists | PMA, IGF-1 |
| Assay Entry | Cell-based terbium phospho-specific activity (stable) |
| Cell State | Dividing Cells |
| Content And Storage | LanthaScreen™ PDCD4 HEK293E cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Gene Symbol | PDCD4 |
| Primary Agonist | Insulin |
| Product Line | LanthaScreen |
| Quantity | 1 Vial |
| Readout | End Point |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.