Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ LanthaScreen™ STAT1 U2OS Cell Line

Catalog No. K1469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
K1469 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. K1469 Supplier Thermo Scientific™ Supplier No. K1469
Only null left
Add to Cart
Add to Cart
Request a Quote

The JAK/STAT1 signaling pathway is known to be activated by type I/II interferons such as interferon-gamma and interferon-alpha.

The JAK/STAT1 signaling pathway is known to be activated by type I/II interferons such as interferon-gamma and interferon-alpha. In this pathway, binding of these cytokines to their respective cell-surface receptors results in the activation of JAKs, which in turn phosphoactivate STAT1 proteins at a specific tyrosine residue (Y701). LanthaScreen™ STAT1 U2OS is a human cell line that constitutively expresses a GFPSTAT1 fusion protein. The JAK/STAT1 signaling pathway is known to be functionally intact in this cell line; therefore, the GFP-STAT1 fusion protein serves as a substrate for the IFN-gamma-inducible phosphorylation by JAKs. Using this cell line, a homogeneous immunoassay has been developed in which the phosphorylation state of GFP-STAT1 is detected in cell lysates using a terbium-labeled anti-pY701-STAT1 antibody in a time-resolved FRET (TR-FRET) readout. The GFP-STAT1 DNA expression construct was transfected into U2OS cells using Lipofectamine™ LTX Reagent, followed by selection with blasticidin.

This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as a sorting marker. Using the lytic TR-FRET immunoassay, this cell line is validated for EC50 and Zfi-factors under optimized conditions using IFN-gamma as a ligand for JAK-mediated GFP-STAT1 phosphorylation. This assay has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance, and assay development time. Additional information using alternate stimuli and alternate assay protocols is available.

TRUSTED_SUSTAINABILITY

Specifications

Alternate Agonists IFN-gamma
Antagonists IFN-alpha
Assay Entry Cell-based terbium phospho-specific activity (stable)
Cell State Dividing Cells
Content And Storage LanthaScreen™ STAT1 U2OS cells are shipped on dry ice.

Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Gene Symbol STAT1
Primary Agonist IFN-γ
Product Line LanthaScreen
Quantity 1 Vial
Readout End Point
Reporter Gene GFP (EmGFP)
Target Entry STAT1
System Type LanthaScreen™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.