Learn More
Thermo Scientific™ LanthaScreen™ STAT3 GripTite™ Cell Line
Description
The JAK/STAT3 signaling pathway is known to be activated by cytokines such as IL-6. In this pathway, binding of IL-6 to its cell-surface receptors results in the activation of JAKs, which in turn phosphoactivate STAT3 proteins at a specific tyrosine residue (Y705). LanthaScreen™ STAT3 GripTite™ is a human cell line that constitutively expresses a GFP-STAT3 fusion protein. The JAK/STAT signaling pathway is known to be functionally intact in this cell line; therefore, the GFP-STAT3 fusion protein serves as a substrate for IL-6-inducible phosphorylation. Using this cell line, a lytic immunoassay has been developed in which the phosphorylation state of GFP-STAT3 is detected in cell lysates using a terbium-labeled anti-pY705-STAT3 antibody in a time-resolved FRET (TR-FRET) readout.
The GFP-STAT3 DNA expression construct was transfected into the GripTite™ HEK 293 cell line using Lipofectamine™ 2000 transfection reagent, and the transfected cells were selected with blasticidin. This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as a sorting marker. The assay utilizing this cell line is validated for EC50 and Z'-factors under optimized conditions using IL-6 as a ligand for JAK-mediated GFP-STAT3 phosphorylation. This assay has also been tested under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance, and assay equilibration time. An alternate 'mix-and-read' assay format is also described.
Specifications
Specifications
| Alternate Agonists | IL-6 |
| Antagonists | IFN-alpha |
| Assay Entry | Cell-based terbium phospho-specific activity (stable) |
| Cell State | Dividing Cells |
| Content And Storage | LanthaScreen™ STAT3 GripTite™ cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Gene Symbol | STAT3 |
| Primary Agonist | IL-6 |
| Product Line | GripTite, LanthaScreen |
| Quantity | 1 Vial |
| Readout | End Point |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.