Learn More
Thermo Scientific™ LanthaScreen™ STAT5 TF-1 Cell Line
Description
LanthaScreen™ GFP Cellular Assays allow for the analysis of post-translational modifications for a number of target proteins in an live-cell context. In hematopoeitic cells, the JAK2/STAT5 signaling pathway plays an essential role in blood cell formation in response to cytokines such as GM-CSF, IL-3, and EPO. In this pathway, binding of these cytokines to their respective cell surface receptors results in the activation of JAK2, which in turn phospho-activates STAT5 proteins at specific tyrosine residues (Tyr-694/699).
LanthaScreen™-STAT5 TF-1 is a human hematopoeitic cell line which constitutively expresses GFP-STAT5 fusion proteins. The JAK2/STAT5 signaling pathway is known to be functionally intact in this cell line, therefore the GFP-STAT5 fusion protein serves as a substrate for the inducible phosphorylation by JAK2. Using this cell line, a homogenous immuno-assay has been developed in which the phosphorylation state of GFP-STAT5 is detected in cell lysates using a LanthaScreen™ Terbium-anti-mouse and anti-phospho STAT5 [pTyr694/699] antibody pair, in a time-resolved FRET (TR-FRET) readout. GFP-STAT5 Lentivirus was transduced into TF-1 cells followed by selection with Blasticidine.
This cell line is a clonal population isolated by flow cytometry using GFP fluorescence as sorting marker, and has been screened for the constitutive expression of GFP-STAT5 fusion protein. Using a lytic TR-FRET immuno-assay, this cell line is validated for EC50 and Z' under optimized conditions using GM-CSF as an agonist for JAK2 -mediated GFP-STAT5 phosphorylation. This assay has also been tested for assay performance under variable experimental conditions, including cell plating density, stimulation time, DMSO tolerance and lysis/equilibration time.
Specifications
Specifications
| Alternate Agonists | IL-3, EPO |
| Antagonists | JAK Inhibitor |
| Assay Entry | Cell-based terbium phospho-specific activity (stable) |
| Cell State | Dividing Cells |
| Content And Storage | LanthaScreen™ STAT5 TF-1 cells are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Detection Method | Fluorescence |
| Gene Symbol | STAT5A |
| Optimal Response | GM-CSF (EC50 = 0.33 ng/mL) |
| Primary Agonist | GM-CSF |
| Product Line | LanthaScreen |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.