Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LanthaScreen™ Tb-peIF2a (pSer52) Antibody

LanthaScreen™ Tb-peIF2a (pSer52) Antibody

$3675.00

Specifications

Antigen Serine/threonine kinase
Concentration 0.47mg/mL
Packaging 1 Each
Primary or Secondary Primary
Storage Requirements –20°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4815
Encompass
View Documents
LIFE TECHNOLOGIES
PV4815
25μg
Each for $3,675.00
Add to cart
 
Description

Description

The alpha subunit of Eukaryotic initiation factor 2 (eIF2alpha) plays a key regulatory role in protein synthesis. In response to a cellular stress, phosphorylation of eIF2alpha at Ser52 occurs by the EIF2AK family of kinases including PERK (EIF2AK3) and PKR (EIF2AK2) to prohibit the initiation of protein synthesis.
For use in combination with the GFP-EIF2alpha physiological protein substrate (PV4809) to evaluate inhibitors in medium to high throughput screening applications.
Specifications

Specifications

Serine/threonine kinase
1 Each
–20°C
Tb (Terbium), lanthanide chelates
RUO
0.47mg/mL
Primary
Biochemical Kinase Assays, Drug Discovery and Development, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation
Liquid
Primary
Safety and Handling

Safety and Handling

SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit