Learn More
Invitrogen™ LanthaScreen™ Tb-pERM (pLRRKtide) Antibody Kit
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants.
Supplier: Invitrogen™ PV4899

Description
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.
For use in combination with the Fluorescein-ERM (LRRKtide) substrate (Cat. No. PV4901) to evaluate inhibitors in medium to high throughput screening applications.

Specifications
Tb (Terbium)/FITC | |
120892 | |
150,000 Da | |
384-well Plate | |
Human | |
End Point | |
LRRK2 D1994A, LRRK2 R1441C, LRRK2 I2020T, LRRK2 G2019S, LRRK2 | |
TR-FRET | |
LanthaScreen™ |
25 μg | |
Terbium-labeled anti-ERM [pLRRKtide] antibody in HEPES buffered saline. Store at -20°C. | |
Biochemical terbium phospho-specific activity | |
340/490, 520 nm | |
Dry Ice | |
Hu | |
Serine/Threonine Kinases | |
LRRK2 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.