Learn More
Invitrogen™ LanthaScreen™ Tb-pERM (pLRRKtide) Antibody Kit
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants.
$6020.00 - $30010.00
Specifications
Antibody Molecular Weight 150,000 Da
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PV4899
Encompass_Preferred
View Documents
Invitrogen™
PV4899
25 μg
Each for $6,020.00
Only null left
Add to Cart
PV4900
Encompass_Preferred
View Documents
Invitrogen™
PV4900
1 mg
Each for $30,010.00
Only null left
Add to Cart
Description
Description
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.
For use in combination with the Fluorescein-ERM (LRRKtide) substrate (Cat. No. PV4901) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
Specifications
150,000 Da
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.