Learn More
Invitrogen™ LanthaScreen™ Tb-pERM (pLRRKtide) Antibody Kit
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants.
Supplier: Invitrogen™ PV4900
Description
A phospho specific antibody directed to the ezrin/radixin/moesin (ERM) peptide that is a substrate for LRRK2 (Leucine-rich repeat kinase 2) and LRRK2 mutants. Mutations in LRRK2 have been found to be related to late-onset Parkinson's disease.
For use in combination with the Fluorescein-ERM (LRRKtide) substrate (Cat. No. PV4901) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
| Serine/threonine kinase | |
| Tb (Terbium)/FITC | |
| 1 mg | |
| Protein Assays and Analysis | |
| 120892 | |
| Terbium-labeled anti-ERM [pLRRKtide] antibody in HEPES buffered saline. Store at -20°C. | |
| 384-well Plate | |
| Human | |
| End Point | |
| Serine/Threonine Kinases | |
| LRRK2 |
| Kinase Assay, Enzyme and Protein Activity Assays, Kinase Assay | |
| AURA17, DARDARIN, PARK8, RIPK7, ROCO | |
| RUO | |
| Primary | |
| 1 Each | |
| 150,000 Da | |
| 340/490, 520 nm | |
| Dry Ice | |
| Hu | |
| TR-FRET | |
| LanthaScreen™ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.