Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP (TGF-beta 1) Antibody (1015022), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB295694 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295694 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295694 Supplier Novus Biologicals Supplier No. FAB77541N

Mouse Monoclonal Antibody

LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, CyTOF

Specifications

Antigen LAP (TGF-beta 1)
Applications ELISA, Flow Cytometry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 1015022
Conjugate Alexa Fluor 700
Dilution ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human LAP (TGF-beta 1), Met1-Ser390, Accession # P01137
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.