Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP (TGF-beta 1) Antibody (7F6), FITC, Novus Biologicals™
SDP

Catalog No. NBP222114F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222114F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222114F Supplier Novus Biologicals Supplier No. NBP222114F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LAP (TGF-beta 1)
Applications Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7F6
Conjugate FITC
Dilution Flow Cytometry, ELISA, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Gene Symbols TGFB1
Host Species Mouse
Immunogen Purified recombinant fragment of human TGF beta 1 (amino acids 62-197) expressed in E. coli. [UniProt# P01137]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.