Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP (TGF-beta 1) Antibody (860206), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB374305 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374305 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374305 Supplier Novus Biologicals Supplier No. FAB7666W
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen LAP (TGF-beta 1)
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 860206
Conjugate DyLight 650
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse LAP (TGF-beta 1), Met1-Ser390, Accession # P04202
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7040
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.