Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP1 Antibody (RL13), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB1202737FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1202737FR 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1202737FR Supplier Novus Biologicals Supplier No. NB1202737FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAP1 Monoclonal specifically detects LAP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LAP1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RL13
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Gene Alias lamina-associated polypeptide 1B, torsin A interacting protein 1
Gene Symbols TOR1AIP1
Host Species Mouse
Immunogen Pore complex-lamina fraction isolated from rat liver nuclear envelopes.
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 208263
Test Specificity Detects lamin associated polypeptide (LAP) 1A, 1B & 1C from rat tissues. This does not cross-react with LAP 2.This detects 3 distinct proteins of molecular weights 75, 68 and 55 kDa representing LAP 1A, 1B and 1C, respectively in rat liver nuclear envelope fractions. Immunofluorescence staining of LAP 1 in rat liver with NB120-2737 results in exclusive labeling of the nuclear periphery and exhibits co-localization with lamin staining.
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.