Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAP3 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB097169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB097169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB097169 Supplier Novus Biologicals Supplier No. NBP297066AF488

Rabbit Polyclonal Antibody

LAP3 Polyclonal specifically detects LAP3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen LAP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias EC 3.4.11, EC 3.4.11.5, LAP, LAPEPEC 3.4.11.1, leucine aminopeptidase 3Prolyl aminopeptidase, Leucyl aminopeptidase, PEPScytosol aminopeptidase, Peptidase SLAP-3, Proline aminopeptidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant LAP3 (NP_056991.2; Met1-Ala519).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51056
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.