Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAR/PTPRF Antibody (S165-38), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP242172FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242172FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242172FR Supplier Novus Biologicals Supplier No. NBP242172FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAR/PTPRF Monoclonal specifically detects LAR/PTPRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen LAR/PTPRF
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S165-38
Conjugate DyLight 680
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Gene Symbols PTPRF
Host Species Mouse
Immunogen Fusion protein amino acids 1315-1607 (cytoplasmic C-terminus) of human LAR. 97% identical in both rat and mouse. >80% identity with PTPRD and PTPRS. >50% identity with PTPRM and PTPRK.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5792
Test Specificity Greater then 80% identity with PTPRD and PTPRS. Greater then 50% identity with PTPRM and PTPRK.Detects approx 85kDa (full length protein is 210 kDa -smaller due to proteolysis into P-subunit containing transmembrane and intracellular domains.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.