Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAR/PTPRF Antibody (W7C6), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB341095 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341095 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341095 Supplier Novus Biologicals Supplier No. NBP290995APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAR/PTPRF Monoclonal antibody specifically detects LAR/PTPRF in Human, Primate samples. It is validated for Flow Cytometry

Specifications

Antigen LAR/PTPRF
Applications Flow Cytometry
Classification Monoclonal
Clone W7C6
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias LCA-homolog, protein tyrosine phosphatase, receptor type, F, receptor type, F polypeptide, receptor-linked protein-tyrosine phosphatase LAR, receptor-type tyrosine-protein phosphatase F
Host Species Mouse
Immunogen WERI-RB-1 retinoblastoma cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 5792
Target Species Human, Primate
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.