Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARGE Antibody (LARGE-02), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB002531 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002531 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002531 Supplier Novus Biologicals Supplier No. NBP262211IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LARGE Monoclonal specifically detects LARGE in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen LARGE
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone LARGE-02
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Acetylglucosaminyltransferase-like 1A, EC 2.4, glycosyltransferase-like protein LARGE1, KIAA0609acetylglucosaminyltransferase-like protein, LARGE1, like-acetylglucosaminyltransferase, like-glycosyltransferase, MDC1D, MDDGA6, MDDGB6
Gene Symbols LARGE
Host Species Mouse
Immunogen Recombinant fragment of human LARGE1 (amino acids 35-142)
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9215
Test Specificity The antibody MEM-65 recognizes an unique epitope of CD2, a 50 kDa glycoprotein present on the human peripheral blood T-lymphocytes and NK cells; also expressed by all thymocytes.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.