Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARGE Antibody, Novus Biologicals™
SDP

Catalog No. NBP149887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149887 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149887 Supplier Novus Biologicals Supplier No. NBP149887
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

LARGE Polyclonal specifically detects LARGE in Human, Mouse, Rat, Bovine samples. It is validated for Peptide ELISA.

Specifications

Antigen LARGE
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_004728.1
Gene Alias Acetylglucosaminyltransferase-like 1A, EC 2.4, glycosyltransferase-like protein LARGE1, KIAA0609acetylglucosaminyltransferase-like protein, LARGE1, like-acetylglucosaminyltransferase, like-glycosyltransferase, MDC1D, MDDGA6, MDDGB6
Gene Symbols LARGE
Host Species Goat
Immunogen Peptide with sequence RYTASSQRERESLE-C corresponding to N-Terminus (near) according to NP_004728.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9215
Test Specificity Reported variants represent identical protein: NP_004728.1, NP_598397.1
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.