Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ UniStore™ Large Reagent Bottles, Narrow Mouth, PP
SDPSCI_ED

Catalog No. S111078 Shop All United Scientific Supplies, Inc Products
Click to view available options
Capacity (Metric):
2 L
4 L
8 L
Quantity:
1 Ea.
6 Pk.

Catalog No. S111078

Supplier: United Scientific™ BNMP8L

Only null left
Add to Cart
Add to Cart

These large capacity polypropylene reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L) and include a lined HDPE cap for a tight seal.

These large capacity reagent bottles feature narrow mouths, integrated shoulder loops (except 2 L) and include a lined HDPE cap for a tight seal.

  • Constructed of polypropylene, the bottles are translucent, pliable, and have excellent strength for extended use.
  • Ideal for packing, shipping and storage purposes.
  • Cap needs to be removed before autoclaving.
  • Manufactured from USP Class VI resin that is FDA-21CFR compliant for food contact and free of heavy metals.
  • Leakproof closure with integrated seal ring.
  • Precision molded for consistent wall thickness with smooth interior and exterior surfaces.

Specifications

Capacity (Metric) 8 L
Capacity (English) 270.51 oz.
Color Translucent White
Diameter (Metric) Outer Neck 50 mm
For Use With (Application) General Lab Use
Material Polypropylene
Quantity 1 Ea.
Sterility Non-sterile
Product Line UniStore
Closure Included Yes
Disposable Reusable
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 420 mm
Product Type Reagent Bottle
Height (English) 16.54 in.
Closure Material Polypropylene
Closure Size 50 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.