Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LARP1 Rabbit anti-Drosophila, Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LARP1 |
---|---|
Concentration | 0.51 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-172-8948
![]() ![]() |
Proteintech Group Inc
137081AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. Associates with the mRNA 5′cap in an MTOR-dependent manner and associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. Associates with actively translating ribosomes via interaction with PABPC1/PABP and stimulates translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. Positively regulates the replication of dengue virus (DENV).Specifications
LARP1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Drosophila, Human, Mouse, Rat | |
Q6PKG0, Q6ZQ58 | |
23367, 303158, 53567, 73158 | |
LARP1 Fusion Protein Ag4645 | |
Primary | |
-20°C | |
LARP1 |
0.51 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0731, La related protein 1, LARP, LARP1 | |
larp, Larp1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title