Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LARP7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109903 Supplier Invitrogen™ Supplier No. PA5109903
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KSTGDPKGFA FVEFETKEQA AKAIEFLNNP PEEAPRKPGI FPKTVKNKPI PALRVVEEKK KKKKKKGRMK KEDNIQAKEE NMDTSNTSIS KMKRSRPTSE GSDIESTEPQ KQCSKKKKKR DRVEASSLPE VRTGKRKRSS SEDAESLAPR SKVKKIIQKD IIKEASEASK ENRDIEISTE EEKDTGDLKD SSLLKTKRKH KKKHKERHKM GEEVIPLRVL S.

LARP7 gene ontology annotations related to this gene include RNA processing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LARP7
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LARP7
Gene Accession No. Q05CL8, Q4G0J3
Gene Alias ALAZS; C330027G06Rik; D3Wsu161e; DKFZP564K112; HDCMA18P; La ribonucleoprotein 7, transcriptional regulator; La ribonucleoprotein domain family member 7; La ribonucleoprotein domain family, member 7; la-related protein 7; la-related protein 7; LOW QUALITY PROTEIN: la-related protein 7; Larp7; PIP7S; P-TEFb-interaction protein for 7SK stability; Unknown (protein for IMAGE:8171460); Unknown (protein for IMAGE:8281412)
Gene Symbols LARP7
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-380 of human LARP7 (NP_057732.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 28036, 51574
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.