Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAS2 Antibody (OTI2B2), FITC, Novus Biologicals™
SDP

Catalog No. NB288149 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB288149 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB288149 Supplier Novus Biologicals Supplier No. NBP272452F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAS2 Monoclonal antibody specifically detects LAS2 in Human samples. It is validated for Western Blot

Specifications

Antigen LAS2
Applications Western Blot
Classification Monoclonal
Clone OTI2B2
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias C18orf54, chromosome 18 open reading frame 54, hypothetical protein LOC162681, LAS2, MGC33382
Host Species Mouse
Immunogen Full length human recombinant protein of human C18orf54(NP_775800) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162681
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.