Learn More
United Scientific Supplies Laser Optics Demonstrator
SDPSCI_ED
Catalog No. s07391 Shop All United Scientific Supplies, Inc Products
Click to view available options
:
Demonstration kit
Allows over 75 experiments in geometric and physical optics.
  • Built-in He-Ne laser
  • Top-quality optical glass components
  • Max. output power: <5mW
  • Wavelength 632 nm
  • Class III laser product
Specifications
Product Type Kit, Laser Optics Demonstration
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.