Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lass5 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB356942 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB356942 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB356942 Supplier Novus Biologicals Supplier No. NBP176964MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lass5 Polyclonal antibody specifically detects Lass5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Lass5
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias CerS5, FLJ25304, LAG1 homolog, ceramide synthase 5, LAG1 longevity assurance homolog 5, LAG1 longevity assurance homolog 5 (S. cerevisiae), MGC45411, Trh4
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide near the amino terminus of the human LASS5. The immunogen is located within the first 50 amino acids of LASS5. Amino Acid Squence: EGPADGYGYPRGRH
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91012
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.