Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Antibody (11B.12), Novus Biologicals™
SDP

Catalog No. NB019433 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019433 0.025 mL
NBP229700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019433 Supplier Novus Biologicals Supplier No. NBP2297000.025ML

Mouse Monoclonal Antibody

LAT Monoclonal specifically detects LAT in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen LAT
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 11B.12
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500
Formulation 0.1M Tris-Glycine (phH 7.4), 0.15M NaCl and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. O43561
Gene Alias 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Gene Symbols LAT
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 31-233 of human LAT, cytoplasmic domain.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27040
Test Specificity LAT 36/38kDa weight may be detected.
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.