Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Antibody (661002), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB376685 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376685 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376685 Supplier Novus Biologicals Supplier No. FAB63341L

Mouse Monoclonal Antibody

LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen LAT
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 661002
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Host Species Mouse
Immunogen E. coli-derived recombinant human LAT, His30-Gln113, Accession # O43561
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27040
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.