Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Antibody (LAT.10-17), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP200289C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200289C 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200289C Supplier Novus Biologicals Supplier No. NBP200289C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAT Monoclonal specifically detects LAT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Cell Surface).

Specifications

Antigen LAT
Applications Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LAT.10-17
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Flow (Cell Surface)
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Gene Symbols LAT
Host Species Mouse
Immunogen The immunogen for this antibody is LAT.
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 27040
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.