Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latent TGF-beta bp1 Antibody (MM0447-5L38), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB344361 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344361 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344361 Supplier Novus Biologicals Supplier No. NBP211783MFV610

Mouse Monoclonal Antibody

Latent TGF-beta bp1 Monoclonal antibody specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Latent TGF-beta bp1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0447-5L38
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1
Host Species Mouse
Immunogen Human recombinant Latent TGF-beta bp1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Ovarian Carcinoma Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4052
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.