Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ LATEX 400nm Black Mini Antibody Conjugation Kit
Shop All Novus Biologicals Products

Click to view available options
Quantity:
16 μg
40 μg
Description
The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.
Kit Contents:
- 1 vial of 10x Quencher buffer
- 1 vial of Resuspension buffer
- 1 vial of Reaction buffer B
- 1 vial of Reaction buffer A
- 4 or 10 Mini vials of 400nm Black LATEX™
Specifications
Specifications
Content And Storage | -20°C |
Quantity | 16 μg |
Target | LATEX™ 400nm Black Mini |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction