Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latexin Antibody (009), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB333447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB333447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB333447 Supplier Novus Biologicals Supplier No. NBP290379MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Latexin Monoclonal antibody specifically detects Latexin in Mouse samples. It is validated for ELISA

Specifications

Antigen Latexin
Applications ELISA
Classification Monoclonal
Clone 009
Conjugate mFluor Violet 610 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse Latexin (Accession#: NP_058033.2, Glu 2-Glu 222)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 56925
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.