Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latexin Antibody (OTI1E10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N272069A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272069A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272069A405 Supplier Novus Biologicals Supplier No. NBP272069AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Latexin Monoclonal specifically detects Latexin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Latexin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI1E10
Conjugate Alexa Fluor 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor
Gene Symbols LXN
Host Species Mouse
Immunogen Full length recombinant protein of human Latexin (NP_064554) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 56925.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.