Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latrophilin 2/LPHN2 Antibody (1030202), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB175924 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175924 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175924 Supplier Novus Biologicals Supplier No. FAB105881W
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Latrophilin 2/LPHN2 Monoclonal specifically detects Latrophilin 2/LPHN2 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen Latrophilin 2/LPHN2
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 1030202
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Calcium-independent alpha-latrotoxin receptor 2, CIRL2, CIRL-2, CL2, KIAA0786, latrophilin 2, Latrophilin homolog 1, latrophilin homolog 2 (cow), latrophilin-2, LEC1LPHH1latrophilin 1, lectomedin-1
Host Species Mouse
Immunogen Human Latrophilin 2/LPHN2 synthetic peptide, Accession # O95490
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 23266
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.