Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LATS2 Antibody (OTI2D3), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271829R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271829R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271829R Supplier Novus Biologicals Supplier No. NBP271829R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LATS2 Monoclonal specifically detects LATS2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen LATS2
Applications Western Blot
Classification Monoclonal
Clone OTI2D3
Conjugate DyLight 550
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.1, FLJ13161, Kinase phosphorylated during mitosis protein, KPM, Large tumor suppressor homolog 2, LATS (large tumor suppressor, Drosophila) homolog 2, LATS, large tumor suppressor, homolog 2 (Drosophila), serine/threonine kinase KPM, Serine/threonine-protein kinase kpm, serine/threonine-protein kinase LATS2, Warts-like kinase
Gene Symbols LATS2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-237 of human LATS2 (NP_055387) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, GPCR, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 26524.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.