Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lavendustin B, 98%, Thermo Scientific™

Catalog No. aaj67142mcr
Click to view available options
Quantity:
1 mg

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125697-91-8
Molecular Formula C21H19NO5
Molecular Weight (g/mol) 365.385
MDL Number MFCD00133446
InChI Key RTYOLBQXFXYMKY-UHFFFAOYSA-N
Synonym lavendustin b, unii-7t53egq52z, benzoic acid,5-bis 2-hydroxyphenyl methyl amino-2-hydroxy, 5-bis 2-hydroxyphenyl methyl amino-2-hydroxybenzoic acid, acmc-20c8gz, 5-bis 2-hydroxybenzyl amino-2-hydroxybenzoic acid, 5-bis 2-hydroxyphenyl methyl amino-2-hydroxy-benzoic acid, benzoic acid, 5-bis 2-hydroxyphenyl methyl amino-2-hydroxy
PubChem CID 3895
IUPAC Name 5-[bis[(2-hydroxyphenyl)methyl]amino]-2-hydroxybenzoic acid
SMILES C1=CC=C(C(=C1)CN(CC2=CC=CC=C2O)C3=CC(=C(C=C3)O)C(=O)O)O

Specifications

Color White
Quantity 1 mg
Sensitivity Light Sensitive
Formula Weight 365.39
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Lavendustin B
storageNote1 -30° to -10°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.