Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Layilin Antibody (20G8), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP242663V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242663V 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242663V Supplier Novus Biologicals Supplier No. NBP242663V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Layilin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 20G8
Conjugate DyLight 405
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ30977, FLJ31092, layilin
Gene Symbols LAYN
Host Species Mouse
Immunogen Recombinant human LAYN (22-235aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.