Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Layilin Antibody (328024), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB176192 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176192 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176192 Supplier Novus Biologicals Supplier No. FAB2637E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Layilin Monoclonal specifically detects Layilin in Human, Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot.

Specifications

Antigen Layilin
Applications CyTOF, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 328024
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ30977, FLJ31092, layilin
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Layilin, Ala22-Glu220, Accession # NP_849156
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.