Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Layilin Antibody (MM0448-4F33), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP211767A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP211767A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP211767A30 Supplier Novus Biologicals Supplier No. NBP211767AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA.

Specifications

Antigen Layilin
Applications Western Blot, ELISA
Classification Monoclonal
Clone MM0448-4F33
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias FLJ30977, FLJ31092, layilin
Gene Symbols LAYN
Host Species Mouse
Immunogen Human recombinant Layilin
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 143903.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.