Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Layilin Antibody (MM0448-4F33), PE, Novus Biologicals™
SDP

Catalog No. NB361736 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361736 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361736 Supplier Novus Biologicals Supplier No. NBP211767PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Western Blot

Specifications

Antigen Layilin
Applications Western Blot
Classification Monoclonal
Clone MM0448-4F33
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias FLJ30977, FLJ31092, layilin
Host Species Mouse
Immunogen Human recombinant Layilin
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.