Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Lazabemide hydrochloride
GREENER_CHOICESDP

Catalog No. 246010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
24-601-0 10 mg
24-605-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 24-601-0 Supplier Tocris Bioscience™ Supplier No. 2460/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective MAO-B inhibitor

Selective, reversible monoamine oxidase B (MAO-B) inhibitor (IC50 values are 0.03 and > 100 μM for MAO-B and MAO-A respectively). Inhibits monoamine uptake at high concentrations (IC50 values are 86, 123 and > 500 μM for noradrenalin, serotonin and dopamine uptake respectively).

Chemical Identifiers

CAS 103878-83-7
Molecular Formula C8H11Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 236.1
Purity >98%
Molecular Weight (g/mol) 236.096
Chemical Name or Material Lazabemide hydrochloride
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.