Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Broth, Miller PROMO
Shop All Fisherbrand Products
Click to view available options
:
LB Broth, Miller; Formulation: 10 g/L Tryptone, 5 g/L Yeast Extract, 10 g/L Sodium Chloride
Description
- Dehydrated

Specifications
Specifications
Content And Storage | 4°C |
Form | Powder |
Grade | Molecular Genetics |
Media Type | LB Broth |
Product Type | Microbiology Media |
Formulation | 10 g/L Tryptone, 5 g/L Yeast Extract, 10 g/L Sodium Chloride |
For Use With (Application) | Preparing Bacterial Nutrient Media |
Packaging Type | Poly Pail |
Quantity | 2000 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction