Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Medium, Miller

Catalog No. p-7076617
Click to view available options
Quantity:
500 mL
1 L

LB Medium, Miller is a bacterial growth medium used for E. coli cultivation for cloning, DNA plasmid production, and production of recombinant proteins.

LB Medium is used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates. Suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production and production of recombinant proteins. Also suitable for selective cultivation when appropriate antibiotics are added.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • LB Medium, Miller is used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates
  • This product is formulated to enhance growth of recombinant E. coli strains by providing exogenous growth factors tryptone and yeast, which the E. coli would otherwise have to synthesize
  • LB Medium, Miller contains high levels of NaCl to allow for osmotic balance
  • It also contains 10g Bacto tryptone, 5g Bacto yeast extract, and 10g NaCl per liter

Application

  • LB Medium, Miller is suitable for non-selective cultivation of E. coli strains for cloning, DNA plasmid production, and production of recombinant proteins
  • It has been used as a culture medium for Escherichia coli, Vibrio natriegens, Pseudomonas aeruginosa, and Bacillus subtilis strains
TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Packaging Type Plastic Bottle
Quantity 500 mL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.