Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LBX1 Antibody, Novus Biologicals™
SDP

Catalog No. NB019489 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019489 0.025 mg
NBP229859 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019489 Supplier Novus Biologicals Supplier No. NBP2298590.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LBX1 Polyclonal specifically detects LBX1 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen LBX1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.1% Sodium Azide
Gene Accession No. P52954
Gene Alias homeobox, HPX6, HPX-6, ladybird homeobox 1, ladybird homeobox homolog 1, ladybird homeobox homolog 1 (Drosophila), Ladybird homeobox protein homolog 1, LBX1Hlady bird-like homeobox, transcription factor LBX1, transcription factor similar to D. melanogaster homeodomain protein lady birdlate
Gene Symbols LBX1
Host Species Rabbit
Immunogen Synthetic peptide.
Molecular Weight of Antigen 30 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10660
Test Specificity Lbx1 (Ladybird). By Western Blot the antibody recognizes a band at ∼30 kDa on mouse spinal cord lysate.
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.